Anti-SPN Rat Monoclonal Antibody [clone: 1B11]

Lieferant: United States Biological
USBIC2397-76REA 432 EUR
USBIC2397-76R
Anti-SPN Rat Monoclonal Antibody [clone: 1B11]
Antikörper
Recommended dilution: Flow Cytometry: 1:5-1:10;

Type: Primary
Antigen: SPN (sialophorin)
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR