Anti-GP1BA Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K236]

Lieferant: United States Biological
USBIC2395-11EEA 0 EUR
USBIC2395-11E
Anti-GP1BA Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K236]
Antikörper
Type: Primary
Antigen: GP1BA (glycoprotein Ib (platelet), alpha polypeptide)
Clonality: Monoclonal
Clone: 5K236
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR