Anti-CD42a,b,c,d Complex Mouse Monoclonal Antibody [clone: 6D267]

Lieferant: United States Biological
USBIC2394-68EA 0 EUR
USBIC2394-68
Anti-CD42a,b,c,d Complex Mouse Monoclonal Antibody [clone: 6D267]
Antikörper
Type: Primary
Antigen: CD42a,b,c,d Complex
Clonality: Monoclonal
Clone: 6D267
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR