Anti-ITGA2B Mouse Monoclonal Antibody [clone: HIP8]

Lieferant: United States Biological
USBIC2394-45EEA 343 EUR
USBIC2394-45E
Anti-ITGA2B Mouse Monoclonal Antibody [clone: HIP8]
Antikörper
Type: Primary
Antigen: ITGA2B (integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigen CD41))
Clonality: Monoclonal
Clone: HIP8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR