Anti-CD40 Rat Monoclonal Antibody [clone: 5K254]

Lieferant: United States Biological
USBIC2392-17AXEA 411 EUR
USBIC2392-17AX
Anti-CD40 Rat Monoclonal Antibody [clone: 5K254]
Antikörper
Type: Primary
Antigen: CD40 (CD40 molecule, TNF receptor superfamily member 5)
Clonality: Monoclonal
Clone: 5K254
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR