Anti-CD40 Mouse Monoclonal Antibody [clone: LOB7/6]

Lieferant: United States Biological
USBIC2392-03REA 0 EUR
USBIC2392-03R
Anti-CD40 Mouse Monoclonal Antibody [clone: LOB7/6]
Antikörper
Recommended dilution: Flow Cytometry: 1:100- 1:200

Type: Primary
Antigen: CD40 (CD40 molecule, TNF receptor superfamily member 5)
Clonality: Monoclonal
Clone: LOB7/6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR