Anti-CD40 Mouse Monoclonal Antibody [clone: 10B1546]

Lieferant: United States Biological
USBIC2392-02PEA 432 EUR
USBIC2392-02P
Anti-CD40 Mouse Monoclonal Antibody [clone: 10B1546]
Antikörper
Recommended dilution: ELISA: 1 ?g/ml; Western Blot: 10 ?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CD40 (CD40 molecule, TNF receptor superfamily member 5)
Clonality: Monoclonal
Clone: 10B1546
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR