Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2]

Lieferant: United States Biological
USBIC2390-05UEA 0 EUR
USBIC2390-05U
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2]
Antikörper
Recommended dilution: Immunochemistry: 40 ?g/ml

Type: Primary
Antigen: CD38 (CD38 molecule)
Clonality: Monoclonal
Clone: HIT2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR