Anti-CD38 Mouse Monoclonal Antibody [clone: 7H157]

Lieferant: United States Biological
USBIC2390-05AEA 473 EUR
USBIC2390-05A
Anti-CD38 Mouse Monoclonal Antibody [clone: 7H157]
Antikörper
Type: Primary
Antigen: CD38 (CD38 molecule)
Clonality: Monoclonal
Clone: 7H157
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR