Anti-CD36 Mouse Monoclonal Antibody [clone: VM58]

Lieferant: United States Biological
USBIC2388-20BEA 530 EUR
USBIC2388-20B
Anti-CD36 Mouse Monoclonal Antibody [clone: VM58]
Antikörper
Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: Monoclonal
Clone: VM58
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR