Anti-CD33 Mouse Monoclonal Antibody [clone: 5K280]

Lieferant: United States Biological
USBIC2385-05CEA 0 EUR
USBIC2385-05C
Anti-CD33 Mouse Monoclonal Antibody [clone: 5K280]
Antikörper
Type: Primary
Antigen: CD33 (CD33 molecule)
Clonality: Monoclonal
Clone: 5K280
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR