Anti-CD33 Mouse Monoclonal Antibody [clone: HIM3-4]

Lieferant: United States Biological
USBIC2385-01NEA 339 EUR
USBIC2385-01N
Anti-CD33 Mouse Monoclonal Antibody [clone: HIM3-4]
Antikörper
Type: Primary
Antigen: CD33 (CD33 molecule)
Clonality: Monoclonal
Clone: HIM3-4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR