Anti-FCGR2/3 Rat monoclonal antibody

Lieferant: United States Biological
USBIC2384-13A-500EA 401 EUR
USBIC2384-13A-500
Anti-FCGR2/3 Rat monoclonal antibody
Antikörper
Type: Primary
Antigen: FCGR2/3 (Fc fragment of IgG, low affinity II/III)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Lambda
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR