Anti-FCGR2B Mouse monoclonal antibody [clone: 2E10]

Lieferant: United States Biological
USBIC2384-03MEA 570 EUR
USBIC2384-03M
Anti-FCGR2B Mouse monoclonal antibody [clone: 2E10]
Antikörper
Recommended dilution: Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: FCGR2B (Fc fragment of IgG, low affinity IIb, receptor (CD32))
Clonality: Monoclonal
Clone: 20000000000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR