Anti-PECAM1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 12K31]

Lieferant: United States Biological
USBIC2383-23GEA 504 EUR
USBIC2383-23G
Anti-PECAM1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 12K31]
Antikörper
Do not freeze!

Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: Monoclonal
Clone: 12K31
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR