Anti-PECAM1 Rat Monoclonal Antibody [clone: 390]

Lieferant: United States Biological
USBIC2383-23AXEA 0 EUR
USBIC2383-23AX
Anti-PECAM1 Rat Monoclonal Antibody [clone: 390]
Antikörper
Type: Primary
Antigen: PECAM1 (platelet/endothelial cell adhesion molecule 1)
Clonality: Monoclonal
Clone: 390
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR