Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 7H141]

Lieferant: United States Biological
USBIC2382-03FEA 473 EUR
USBIC2382-03F
Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 7H141]
Antikörper
Type: Primary
Antigen: TNFRSF8 (tumor necrosis factor receptor superfamily, member 8)
Clonality: Monoclonal
Clone: 7H141
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR