Anti-CD28 Hamster Monoclonal Antibody (Biotin) [clone: 37.51]

Lieferant: United States Biological
USBIC2380-18CEA 394 EUR
USBIC2380-18C
Anti-CD28 Hamster Monoclonal Antibody (Biotin) [clone: 37.51]
Antikörper
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: Monoclonal
Clone: 37.51
Conjugation: Biotin
Epitope:
Host: Hamster
Isotype: IgG Lambda
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR