Anti-CD28 Mouse Monoclonal Antibody [clone: E18]

Lieferant: United States Biological
USBIC2380-01FEA 752 EUR
USBIC2380-01F
Anti-CD28 Mouse Monoclonal Antibody [clone: E18]
Antikörper
Recommended dilution: Flow Cytometry: 1:50-1:100;

Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: Monoclonal
Clone: E18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR