Anti-IL2RA Mouse Monoclonal Antibody (Biotin) [clone: 5G44]

Lieferant: United States Biological
USBIC2340-01EA 357 EUR
USBIC2340-01
Anti-IL2RA Mouse Monoclonal Antibody (Biotin) [clone: 5G44]
Antikörper
Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: Monoclonal
Clone: 5G44
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR