Anti-IL2RA Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A262]

Lieferant: United States Biological
USBIC2276-01CEA 604 EUR
USBIC2276-01C
Anti-IL2RA Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A262]
Antikörper
Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: Monoclonal
Clone: 6A262
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR