Anti-CD24 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K251]

Lieferant: United States Biological
USBIC2275-11HEA 292 EUR
USBIC2275-11H
Anti-CD24 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K251]
Antikörper
Type: Primary
Antigen: CD24 (CD24 molecule)
Clonality: Monoclonal
Clone: 5K251
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2c Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR