Anti-CD24 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K10]

Lieferant: United States Biological
USBIC2275-05DEA 324 EUR
USBIC2275-05D
Anti-CD24 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K10]
Antikörper
Do not freeze!

Type: Primary
Antigen: CD24 (CD24 molecule)
Clonality: Monoclonal
Clone: 5K10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR