Anti-FCER2 Rat monoclonal antibody [clone: 5K250]

Lieferant: United States Biological
USBIC2273-91AXEA 481 EUR
USBIC2273-91AX
Anti-FCER2 Rat monoclonal antibody [clone: 5K250]
Antikörper
Type: Primary
Antigen: FCER2 (Fc fragment of IgE, low affinity II, receptor for (CD23))
Clonality: Monoclonal
Clone: 5K250
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR