Anti-CD22 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC2273-17BEA 0 EUR
USBIC2273-17B
Anti-CD22 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 0.5-1 ?g/ml; Western Blot: 0.1-0.5 ?g/ml

Type: Primary
Antigen: CD22 (CD22 molecule)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR