Anti-CD22 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3H1116]

Lieferant: United States Biological
USBIC2273-02F2EA 553 EUR
USBIC2273-02F2
Anti-CD22 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3H1116]
Antikörper
Recommended dilution: Flow Cytometry:1:5;
Do not freeze!

Type: Primary
Antigen: CD22 (CD22 molecule)
Clonality: Monoclonal
Clone: 3H1116
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR