Anti-CR2 Mouse Monoclonal Antibody (Biotin) [clone: 7H167]

Lieferant: United States Biological
USBIC2272-56BEA 414 EUR
USBIC2272-56B
Anti-CR2 Mouse Monoclonal Antibody (Biotin) [clone: 7H167]
Antikörper
Type: Primary
Antigen: CR2 (complement component (3d/Epstein Barr virus) receptor 2)
Clonality: Monoclonal
Clone: 7H167
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR