Anti-MS4A1 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIC2270-05HEA 471 EUR
USBIC2270-05H
Anti-MS4A1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR