Anti-MS4A1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K57]

Lieferant: United States Biological
USBIC2270-04CEA 467 EUR
USBIC2270-04C
Anti-MS4A1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K57]
Antikörper
Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: Monoclonal
Clone: 5K57
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR