Anti-MS4A1 Mouse Monoclonal Antibody [clone: 9A138]

Lieferant: United States Biological
USBIC2270-02AEA 431 EUR
USBIC2270-02A
Anti-MS4A1 Mouse Monoclonal Antibody [clone: 9A138]
Antikörper
Recommended dilution: Flow Cytometry: 1:50-1:100;

Type: Primary
Antigen: MS4A1 (membrane-spanning 4-domains, subfamily A, member 1)
Clonality: Monoclonal
Clone: 9A138
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR