Anti-CD19 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin) [clone: 10B936/3H2592]

Lieferant: United States Biological
USBIC2269-98EA 561 EUR
USBIC2269-98
Anti-CD19 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin) [clone: 10B936/3H2592]
Antikörper
Do not freeze!

Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: Monoclonal
Clone: 10B936/3H2592
Conjugation: FITC (Fluorescein)/PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mixed
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR