Anti-CD19 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin) [clone: 5F22/5F7]
Lieferant: United States Biological
USBIC2269-94EA 540 EUR
USBIC2269-94
Anti-CD19 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin) [clone: 5F22/5F7]
Antikörper
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: Monoclonal
Clone: 5F22/5F7
Conjugation: FITC (Fluorescein)/PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1+IgG1
Reactivity: Human