Anti-CD19 Mouse Monoclonal Antibody [clone: 5K218]

Lieferant: United States Biological
USBIC2269-83CEA 261 EUR
USBIC2269-83C
Anti-CD19 Mouse Monoclonal Antibody [clone: 5K218]
Antikörper
Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: Monoclonal
Clone: 5K218
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR