Anti-CD19 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6D5]

Lieferant: United States Biological
USBIC2269-50MEA 319 EUR
USBIC2269-50M
Anti-CD19 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 6D5]
Antikörper
Do not freeze!

Type: Primary
Antigen: CD19 (CD19 molecule)
Clonality: Monoclonal
Clone: 6D5
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR