Anti-ITGB2 Mouse Monoclonal Antibody [clone: 7H193]

Lieferant: United States Biological
USBIC2269-01EXEA 1110 EUR
USBIC2269-01EX
Anti-ITGB2 Mouse Monoclonal Antibody [clone: 7H193]
Antikörper
Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: Monoclonal
Clone: 7H193
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR