Anti-FCGR3A Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 10K42]

Lieferant: United States Biological
USBIC2267-60AEA 565 EUR
USBIC2267-60A
Anti-FCGR3A Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 10K42]
Antikörper
Do not freeze!

Type: Primary
Antigen: FCGR3A (Fc fragment of IgG, low affinity IIIa, receptor (CD16a))
Clonality: Monoclonal
Clone: 10K42
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR