Anti-FUT4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5F17]

Lieferant: United States Biological
USBIC2266-06BEA 418 EUR
USBIC2266-06B
Anti-FUT4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5F17]
Antikörper
Type: Primary
Antigen: FUT4 (fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific))
Clonality: Monoclonal
Clone: 5F17
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR