Anti-CD14 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)/Cy5®-PE (Phycoerythrin) [clone: 5F16/5F14/5F41]

Lieferant: United States Biological
USBIC2265-90EA 506 EUR
USBIC2265-90
Anti-CD14 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)/Cy5®-PE (Phycoerythrin) [clone: 5F16/5F14/5F41]
Antikörper
Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: Monoclonal
Clone: 5F16/5F14/5F41
Conjugation: FITC (Fluorescein)/PE (Phycoerythrin)/Cy5®-PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1+IgG1+IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR