Anti-CD14 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5F16]

Lieferant: United States Biological
USBIC2265-33KEA 345 EUR
USBIC2265-33K
Anti-CD14 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5F16]
Antikörper
Type: Primary
Antigen: CD14 (CD14 molecule)
Clonality: Monoclonal
Clone: 5F16
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR