Anti-ANPEP Rat Monoclonal Antibody [clone: 9G140]

Lieferant: United States Biological
USBIC2264-10DEA 515 EUR
USBIC2264-10D
Anti-ANPEP Rat Monoclonal Antibody [clone: 9G140]
Antikörper
Recommended dilution: Flow Cytometry: 1:50; Immunochemistry: 1:50

Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: Monoclonal
Clone: 9G140
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR