Anti-ANPEP Mouse Monoclonal Antibody [clone: 3H1013]

Lieferant: United States Biological
USBIC2264-03LEA 752 EUR
USBIC2264-03L
Anti-ANPEP Mouse Monoclonal Antibody [clone: 3H1013]
Antikörper
Recommended dilution: Flow Cytometry: 1:50-1:100;

Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: Monoclonal
Clone: 3H1013
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR