Anti-ANPEP Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3H1010]

Lieferant: United States Biological
USBIC2264-03KEA 621 EUR
USBIC2264-03K
Anti-ANPEP Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3H1010]
Antikörper
Do not freeze!

Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: Monoclonal
Clone: 3H1010
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR