Anti-ANPEP Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5F14]

Lieferant: United States Biological
USBIC2264-02AEA 382 EUR
USBIC2264-02A
Anti-ANPEP Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5F14]
Antikörper
Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: Monoclonal
Clone: 5F14
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR