Anti-CD13, Triple Color Mouse Monoclonal Antibody [clone: Tuk 1]

Lieferant: United States Biological
USBIC2264-01MEA 747 EUR
USBIC2264-01M
Anti-CD13, Triple Color Mouse Monoclonal Antibody [clone: Tuk 1]
Antikörper
Type: Primary
Antigen: CD13, Triple Color
Clonality: Monoclonal
Clone: Tuk 1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR