Anti-ANPEP Mouse Monoclonal Antibody [clone: 7H112]

Lieferant: United States Biological
USBIC2264-01AEA 473 EUR
USBIC2264-01A
Anti-ANPEP Mouse Monoclonal Antibody [clone: 7H112]
Antikörper
Type: Primary
Antigen: ANPEP (alanyl (membrane) aminopeptidase)
Clonality: Monoclonal
Clone: 7H112
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR