Anti-ITGAX Mouse Monoclonal Antibody [clone: 10B633]

Lieferant: United States Biological
USBIC2262-56TEA 573 EUR
USBIC2262-56T
Anti-ITGAX Mouse Monoclonal Antibody [clone: 10B633]
Antikörper
Recommended dilution: Flow Cytometry:

Type: Primary
Antigen: ITGAX (integrin, alpha X (complement component 3 receptor 4 subunit))
Clonality: Monoclonal
Clone: 10B633
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR