Anti-ITGAM Rat Monoclonal Antibody

Lieferant: United States Biological
USBIC2262-43K-500EA 473 EUR
USBIC2262-43K-500
Anti-ITGAM Rat Monoclonal Antibody
Antikörper
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Human, Monkey, Mouse, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR