Anti-ITGAM Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 5K275]

Lieferant: United States Biological
USBIC2262-36DEA 411 EUR
USBIC2262-36D
Anti-ITGAM Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy5®) [clone: 5K275]
Antikörper
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: Monoclonal
Clone: 5K275
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR