Anti-ITGAL Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B581]

Lieferant: United States Biological
USBIC2262-06PEA 0 EUR
USBIC2262-06P
Anti-ITGAL Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 10B581]
Antikörper
Type: Primary
Antigen: ITGAL (integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide))
Clonality: Monoclonal
Clone: 10B581
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Pig, Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR