Anti-ITGAL Goat Polyclonal Antibody

Lieferant: United States Biological
USBIC2262-02A1EA 633 EUR
USBIC2262-02A1
Anti-ITGAL Goat Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-2?g/ml; ELISA: 1:16000; Immunochemistry: 3. 75 ?g/ml

Type: Primary
Antigen: ITGAL (integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen 1; alpha polypeptide))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR